siRNA Details
| Gene Symbol: Mink1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001045959.1 | NP_001039424.1 | 27 | 3448 | ||
| NM_001045964.1 | NP_001039429.1 | 28 | 3469 | |||
| NM_016713.2 | NP_057922.2 | 26 | 3337 | |||
| NM_176893.2 | NP_795712.2 | 27 | 3361 | |||
| GenBank mRNA | AB035697.1 | - | 26 | 3322 | ||
| AB041925.1 | - | 27 | 3347 | |||
| AK220178.1 | - | 9 | 887 | |||
| BC011346.1 | - | 23 | 2841 | |||
| BC052474.1 | - | 27 | 3303 | 6833893 | ||
| BC079549.1 | - | 26 | 3819 | |||

Mink1
Genomic Map