siRNA Details
| Gene Symbol: H2afz | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016750.3 | NP_058030.1 | 5 | 813 | ||
| GenBank mRNA | AK028176.1 | - | 5 | 710 | ||
| AK088704.1 | - | 5 | 760 | |||
| AK143572.1 | - | 5 | 712 | |||
| AK150454.1 | - | 5 | 712 | |||
| AK151052.1 | - | 5 | 712 | |||
| AK151509.1 | - | 5 | 712 | |||
| AK151868.1 | - | 5 | 712 | |||
| AK152398.1 | - | 5 | 710 | |||
| AK152848.1 | - | 5 | 711 | |||
| AK168178.1 | - | 5 | 712 | |||
| AK169003.1 | - | 5 | 598 | |||
| AK213534.1 | - | 1 | 103 | |||
| AY074806.1 | - | 5 | 710 | |||
| BC079903.1 | - | 5 | 735 | 30622912 | ||
| U70494.1 | - | 5 | 701 | |||
| Gene Symbol: Gm8203 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK210116.1 | - | 1 | 247 | ||

H2afz
Genomic Map