siRNA Details
| Gene Symbol: Ddah2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001190449.1 | NP_001177378.1 | 2, 3 | 605 | ||
| NM_016765.3 | NP_058045.1 | 2, 3 | 605 | |||
| GenBank mRNA | AF004106.1 | - | 2, 3 | 539 | ||
| BC003328.1 | - | 2, 3 | 555 | 3158175 | ||

Ddah2
Genomic Map