siRNA Details
| Gene Symbol: Bok | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016778.2 | NP_058058.1 | 3, 4 | 536 | ||
| GenBank mRNA | AF027707.1 | - | 3, 4 | 534 | ||
| AK156243.1 | - | 3, 4 | 571 | |||
| AK168373.1 | - | 3, 4 | 467 | |||
| AK171258.1 | - | 3, 4 | 493 | |||
| BC030069.1 | - | 3, 4 | 474 | 2936938 | ||

Bok
Genomic Map