siRNA Details
| Gene Symbol: Fubp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_057172.3 | NP_476513.2 | 8, 9 | 764 | ||
| GenBank mRNA | AK028418.1 | - | 8, 9 | 765 | ||
| AK029458.1 | - | 9, 10 | 783 | |||
| AK032517.1 | - | 8, 9 | 731 | |||
| AK138011.1 | - | 8, 9 | 709 | |||
| AK160815.1 | - | 8, 9 | 702 | |||
| AK166066.1 | - | 8, 9 | 709 | |||
| AK167730.1 | - | 8, 9 | 744 | |||
| BC014763.1 | - | 8, 9 | 727 | 4511225 | ||
| BC023780.1 | - | 8, 9 | 745 | 5347067 | ||

Fubp1
Genomic Map