siRNA Details
| Gene Symbol: Fubp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_057172.3 | NP_476513.2 | 7 | 561 | ||
| GenBank mRNA | AK028418.1 | - | 7 | 562 | ||
| AK029458.1 | - | 8 | 580 | |||
| AK032517.1 | - | 7 | 528 | |||
| AK079204.1 | - | 6 | 2029 | |||
| AK138011.1 | - | 7 | 506 | |||
| AK160815.1 | - | 7 | 499 | |||
| AK166066.1 | - | 7 | 506 | |||
| AK167730.1 | - | 7 | 541 | |||
| BC014763.1 | - | 7 | 524 | 4511225 | ||
| BC023780.1 | - | 7 | 542 | 5347067 | ||

Fubp1
Genomic Map