siRNA Details
| Gene Symbol: Fubp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_057172.3 | NP_476513.2 | 17, 18 | 1826 | ||
| GenBank mRNA | AK028418.1 | - | 17, 18 | 1828 | ||
| AK138011.1 | - | 17, 18 | 1771 | |||
| AK160815.1 | - | 17, 18 | 1764 | |||
| AK166066.1 | - | 17, 18 | 1771 | |||
| AK167730.1 | - | 17, 18 | 1806 | |||

Fubp1
Genomic Map