siRNA Details
| Gene Symbol: Rnf24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178607.4 | NP_848722.1 | 3, 4 | 351 | ||
| GenBank mRNA | AK015699.1 | - | 3, 4 | 322 | ||
| AK048813.1 | - | 3, 4 | 309 | |||
| AK083665.1 | - | 3, 4 | 341 | |||
| AK089207.1 | - | 3, 4 | 338 | |||
| AK154093.1 | - | 3, 4 | 288 | |||
| AK165427.1 | - | 3, 4 | 309 | |||
| BC096529.1 | - | 3, 4 | 284 | 6406985 | ||

Rnf24
Genomic Map