siRNA Details
| Gene Symbol: R3hcc1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177464.4 | NP_803415.1 | 2 | 1403 | ||
| GenBank mRNA | AK082926.1 | - | 2 | 1404 | ||
| AK147893.1 | - | 2 | 1387 | |||
| BC038935.1 | - | 2 | 1379 | 5035077 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

R3hcc1l| Gene Symbol: R3hcc1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177464.4 | NP_803415.1 | 2 | 1403 | ||
| GenBank mRNA | AK082926.1 | - | 2 | 1404 | ||
| AK147893.1 | - | 2 | 1387 | |||
| BC038935.1 | - | 2 | 1379 | 5035077 | ||