siRNA Details
| Gene Symbol: Nus1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030250.2 | NP_084526.1 | 3, 4 | 895 | ||
| GenBank mRNA | AK155620.1 | - | 3, 4 | 896 | ||
| AK162260.1 | - | 1, 2 | 57 | |||
| AK167741.1 | - | 3, 4 | 884 | |||
| AK167939.1 | - | 3, 4 | 888 | |||
| AK214556.1 | - | 1, 2 | 96 | |||
| BC003223.1 | - | 3, 4 | 854 | 3482163 | ||
| BC018372.1 | - | 3, 4 | 870 | 3499868 | ||

Nus1
Genomic Map