siRNA Details
| Gene Symbol: Nus1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030250.2 | NP_084526.1 | 4 | 924 | ||
| GenBank mRNA | AK155620.1 | - | 4 | 925 | ||
| AK162260.1 | - | 2 | 86 | |||
| AK167741.1 | - | 4 | 913 | |||
| AK167939.1 | - | 4 | 917 | |||
| BC003223.1 | - | 4 | 883 | 3482163 | ||
| BC018372.1 | - | 4 | 899 | 3499868 | ||

Nus1
Genomic Map