siRNA Details
| Gene Symbol: Umodl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177465.4 | NP_803416.2 | 11 | 1915 | ||
| GenBank mRNA | AK040446.1 | - | 3 | 416 | ||
| AY771616.1 | - | 11 | 1919 | |||
| AY771617.1 | - | 11 | 1919 | |||
| AY771618.1 | - | 11 | 1919 | |||
| BC051427.1 | - | 11 | 1915 | |||

Umodl1
Genomic Map