siRNA Details
| Gene Symbol: Ppp6r3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001164159.1 | NP_001157631.1 | 3 | 418 | ||
| NM_028999.1 | NP_083275.1 | 4 | 566 | |||
| NM_029456.2 | NP_083732.1 | 4 | 566 | |||
| GenBank mRNA | AK015925.1 | - | 4 | 547 | ||
| AK018652.1 | - | 3 | 407 | |||
| AK035236.1 | - | 4 | 546 | |||
| AK088526.1 | - | 3 | 386 | |||
| AK129392.1 | - | 4 | 508 | |||
| BC008529.1 | - | 4 | 549 | 3601021 | ||
| BY720838.1 | - | 3 | 404 | |||

Ppp6r3
Genomic Map