siRNA Details
| Gene Symbol: Coq5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026504.2 | NP_080780.1 | 3 | 377 | ||
| GenBank mRNA | AK007496.1 | - | 3 | 378 | ||
| AK009825.1 | - | 3 | 373 | |||
| AK014348.1 | - | 3 | 389 | |||
| AK045993.1 | - | 3 | 384 | |||
| AK050334.1 | - | 3 | 384 | |||
| AK217127.1 | - | 2 | 146 | |||
| BC052345.1 | - | 3 | 384 | 6585618 | ||

Coq5
Genomic Map