siRNA Details
| Gene Symbol: Hgsnat | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029884.1 | NP_084160.1 | 6 | 684 | ||
| GenBank mRNA | AK035264.1 | - | 6 | 617 | ||
| AK149883.1 | - | 6 | 688 | |||
| AK152926.1 | - | 6 | 649 | |||
| AK159649.1 | - | 6 | 613 | |||
| AK160068.1 | - | 6 | 607 | |||
| AK191575.1 | - | 382 | ||||
| BC024084.1 | - | 6 | 591 | 5101064 | ||

Hgsnat
Genomic Map