siRNA Details
| Gene Symbol: Hgsnat | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029884.1 | NP_084160.1 | 10 | 1072 | ||
| GenBank mRNA | AK035264.1 | - | 10 | 1005 | ||
| AK149883.1 | - | 10 | 1076 | |||
| AK152926.1 | - | 10 | 1037 | |||
| AK159649.1 | - | 10 | 1001 | |||
| AK160068.1 | - | 10 | 995 | |||
| AK209791.1 | - | 4 | 338 | |||
| BC024084.1 | - | 10 | 979 | 5101064 | ||

Hgsnat
Genomic Map