siRNA Details
| Gene Symbol: Rragd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027491.2 | NP_081767.2 | 2, 3 | 911 | ||
| GenBank mRNA | AF490406.1 | - | 3, 4 | 909 | ||
| AK017818.1 | - | 1, 2 | 271 | |||
| BC038137.1 | - | 2, 3 | 400 | |||
| CK387674.1 | - | 2, 3 | 490 | |||

Rragd
Genomic Map