siRNA Details
| Gene Symbol: Cdca8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026560.4 | NP_080836.3 | 6 | 720 | ||
| GenBank mRNA | AK076065.1 | - | 6 | 677 | ||
| AK076422.1 | - | 6 | 710 | |||
| AK077717.1 | - | 6 | 673 | |||
| AK077920.1 | - | 6 | 643 | |||
| AK078404.1 | - | 6 | 673 | |||
| AK082800.1 | - | 6 | 685 | |||
| AY550907.1 | - | 6 | 710 | |||
| BC068181.1 | - | 5 | 634 | 30634239 | ||

Cdca8
Genomic Map