siRNA Details
| Gene Symbol: D6Ertd527e | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001167937.1 | NP_001161409.1 | 2 | 2154 | ||
| NM_001167938.1 | NP_001161410.1 | 2 | 2151 | |||
| GenBank mRNA | AK139701.1 | - | 2 | 2159 | ||
| AK139822.1 | - | 2 | 2153 | |||

D6Ertd527e
Genomic Map