siRNA Details
| Gene Symbol: Echdc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001254754.1 | NP_001241683.1 | 3, 4 | 403 | ||
| NM_026728.4 | NP_081004.2 | 4, 5 | 471 | |||
| GenBank mRNA | AK005030.1 | - | 4, 5 | 412 | ||
| AK011365.1 | - | 3, 4 | 349 | |||
| AK160482.1 | - | 2, 3 | 174 | |||
| AK166965.1 | - | 4, 5 | 413 | |||

Echdc2
Genomic Map