siRNA Details
| Gene Symbol: Echdc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001254754.1 | NP_001241683.1 | 8, 9 | 829 | ||
| NM_026728.4 | NP_081004.2 | 9, 10 | 897 | |||
| GenBank mRNA | AK005030.1 | - | 9, 10 | 838 | ||
| AK011365.1 | - | 8, 9 | 775 | |||
| AK153878.1 | - | 9, 10 | 872 | |||
| AK160482.1 | - | 7, 8 | 600 | |||
| AK166388.1 | - | 9, 10 | 869 | |||
| AK166965.1 | - | 9, 10 | 839 | |||
| BC025104.1 | - | 9, 10 | 843 | 3591648 | ||
| BC025454.1 | - | 5, 6 | 381 | 5352625 | ||

Echdc2
Genomic Map