siRNA Details
| Gene Symbol: Echdc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001254754.1 | NP_001241683.1 | 6 | 652 | ||
| NM_026728.4 | NP_081004.2 | 7 | 720 | |||
| GenBank mRNA | AK005030.1 | - | 7 | 661 | ||
| AK011365.1 | - | 6 | 598 | |||
| AK153878.1 | - | 7 | 695 | |||
| AK160482.1 | - | 5 | 423 | |||
| AK166388.1 | - | 7 | 692 | |||
| AK166965.1 | - | 7 | 662 | |||
| BC025104.1 | - | 7 | 666 | 3591648 | ||
| BC025454.1 | - | 3 | 204 | 5352625 | ||

Echdc2
Genomic Map