siRNA Details
| Gene Symbol: Ppp2r2d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026391.2 | NP_080667.1 | 6 | 645 | ||
| GenBank mRNA | AF366393.1 | - | 6 | 553 | ||
| AK005033.1 | - | 5 | 534 | |||
| AK011198.1 | - | 6 | 667 | |||
| AK129386.2 | - | 6 | 672 | |||
| BC066022.1 | - | 6 | 645 | 6828707 | ||

Ppp2r2d
Genomic Map