siRNA Details
| Gene Symbol: Tax1bp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025816.3 | NP_080092.2 | 18 | 2429 | ||
| GenBank mRNA | AK004574.1 | - | 18 | 2411 | ||
| AK146076.1 | - | 18 | 2409 | |||
| BC014798.1 | - | 18 | 2350 | 3962810 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tax1bp1| Gene Symbol: Tax1bp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025816.3 | NP_080092.2 | 18 | 2429 | ||
| GenBank mRNA | AK004574.1 | - | 18 | 2411 | ||
| AK146076.1 | - | 18 | 2409 | |||
| BC014798.1 | - | 18 | 2350 | 3962810 | ||