siRNA Details
| Gene Symbol: Cenpo | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134046.5 | NP_598807.2 | 5 | 1168 | ||
| GenBank mRNA | AK013198.1 | - | 5 | 840 | ||
| AK018444.1 | - | 5 | 813 | |||
| AK029226.1 | - | 5 | 1169 | |||
| AK036089.1 | - | 4 | 673 | |||
| AK134266.1 | - | 5 | 840 | |||
| BC034399.1 | - | 5 | 827 | 4481047 | ||

Cenpo
Genomic Map