siRNA Details
| Gene Symbol: Zfp622 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_144523.2 | NP_653106.1 | 3 | 1220 | ||
| GenBank mRNA | AK031232.1 | - | 3 | 1220 | ||
| AK149106.1 | - | 3 | 1151 | |||
| AK154951.1 | - | 3 | 1146 | |||
| AK170489.1 | - | 2 | 1333 | |||
| BC006964.1 | - | 3 | 1125 | 3153893 | ||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: NM_001009652, XM_008760822, BC088214, AY321329, AY310142.

Zfp622
Genomic Map