siRNA Details
| Gene Symbol: Tspan14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145928.1 | NP_666040.1 | 7, 8 | 671 | ||
| GenBank mRNA | AK030593.1 | - | 7, 8 | 673 | ||
| AK156671.1 | - | 7, 8 | 684 | |||
| BC024611.1 | - | 7, 8 | 676 | 4188261 | ||
| BC025568.1 | - | 7, 8 | 685 | 5322531 | ||
| BC026574.1 | - | 7, 8 | 671 | 4954874 | ||

Tspan14
Genomic Map