siRNA Details
| Gene Symbol: Ncald | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001170866.1 | NP_001164337.1 | 5 | 458 | ||
| NM_001170867.1 | NP_001164338.1 | 3 | 392 | |||
| NM_001170868.1 | NP_001164339.1 | 2 | 340 | |||
| NM_134094.4 | NP_598855.2 | 5 | 648 | |||
| GenBank mRNA | AI893360.1 | - | 5 | 454 | ||
| AK034031.1 | - | 3 | 371 | |||
| AK048440.1 | - | 5 | 628 | |||
| AK167313.1 | - | 2 | 310 | |||
| BC011162.1 | - | 2 | 335 | 4191195 | ||
| BC026979.1 | - | 2 | 294 | 5365560 | ||

Ncald
Genomic Map