siRNA Details
| Gene Symbol: Nit2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023175.1 | NP_075664.1 | 4 | 359 | ||
| GenBank mRNA | AF284573.1 | - | 4 | 359 | ||
| AK003604.1 | - | 1 | 54 | |||
| AK004533.1 | - | 4 | 343 | |||
| AK004535.1 | - | 4 | 345 | |||
| BC020153.1 | - | 4 | 348 | 3992930 | ||

Nit2
Genomic Map