siRNA Details
| Gene Symbol: Cisd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134007.4 | NP_598768.1 | 2 | 201 | ||
| GenBank mRNA | AK075907.1 | - | 2 | 202 | ||
| AK131595.1 | - | 2 | 201 | |||
| AK131605.1 | - | 2 | 180 | |||
| AK131812.1 | - | 2 | 205 | |||
| AK131995.1 | - | 2 | 212 | |||
| AK167898.1 | - | 2 | 180 | |||
| BC013522.1 | - | 2 | 190 | 4236502 | ||
| BC019860.1 | - | 2 | 217 | 5123692 | ||
| BC021952.1 | - | 2 | 238 | 5102315 | ||

Cisd1
Genomic Map