siRNA Details
| Gene Symbol: Wipi1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145940.2 | NP_666052.1 | 11 | 1162 | ||
| GenBank mRNA | AK031672.1 | - | 11 | 1171 | ||
| AK034907.1 | - | 11 | 1206 | |||
| AK050495.1 | - | 11 | 1170 | |||
| AK159608.1 | - | 11 | 1191 | |||
| AK170563.1 | - | 11 | 1206 | |||
| BC024811.1 | - | 11 | 1051 | |||
| BC024883.1 | - | 11 | 1126 | |||
| BC025560.1 | - | 11 | 1162 | 5322999 | ||

Wipi1
Genomic Map