siRNA Details
| Gene Symbol: D13Ertd608e | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK143258.1 | - | 2 | 408 | ||
| BC147106.1 | - | 2 | 403 | 9056131 | ||
| BC147107.1 | - | 2 | 403 | 9056120 | ||
| BX525838.1 | - | 2 | 392 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

D13Ertd608e| Gene Symbol: D13Ertd608e | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK143258.1 | - | 2 | 408 | ||
| BC147106.1 | - | 2 | 403 | 9056131 | ||
| BC147107.1 | - | 2 | 403 | 9056120 | ||
| BX525838.1 | - | 2 | 392 | |||