siRNA Details
| Gene Symbol: Setd3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028262.3 | NP_082538.2 | 6 | 795 | ||
| GenBank mRNA | AK011993.1 | - | 6 | 775 | ||
| AK029403.1 | - | 5 | 626 | |||
| AK031371.1 | - | 6 | 758 | |||
| AK146777.1 | - | 6 | 789 | |||
| AK166570.1 | - | 6 | 727 | |||
| AK194709.1 | - | 1 | 253 | |||
| AK208208.1 | - | 1 | 231 | |||
| AY513271.1 | - | 6 | 728 | |||
| BC016123.1 | - | 6 | 725 | 4503842 | ||
| BC019973.1 | - | 6 | 725 | 4195130 | ||
| BC057968.1 | - | 5 | 589 | 4979330 | ||
| BC108358.1 | - | 6 | 766 | |||

Setd3
Genomic Map