siRNA Details
| Gene Symbol: Krr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178610.4 | NP_848725.2 | 9, 10 | 1106 | ||
| GenBank mRNA | AK011972.1 | - | 9, 10 | 1016 | ||
| AK012108.1 | - | 9, 10 | 1044 | |||
| AK030560.1 | - | 9, 10 | 1018 | |||
| AK032472.1 | - | 9, 10 | 1018 | |||
| AK036447.1 | - | 8, 9 | 1104 | |||
| AK041854.1 | - | 9, 10 | 1018 | |||
| AK078627.1 | - | 3, 4 | 262 | |||
| AK146926.1 | - | 9, 10 | 1023 | |||
| AK168675.1 | - | 9, 10 | 1018 | |||
| BC089510.1 | - | 9, 10 | 1023 | 30246541 | ||
| BC094236.1 | - | 9, 10 | 1007 | |||

Krr1
Genomic Map