siRNA Details
| Gene Symbol: Krr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178610.4 | NP_848725.2 | 7 | 800 | ||
| GenBank mRNA | AK012108.1 | - | 7 | 738 | ||
| AK030560.1 | - | 7 | 712 | |||
| AK032472.1 | - | 7 | 712 | |||
| AK036447.1 | - | 6 | 798 | |||
| AK041854.1 | - | 7 | 712 | |||
| AK146926.1 | - | 7 | 718 | |||
| AK168675.1 | - | 7 | 712 | |||
| BC089510.1 | - | 7 | 717 | 30246541 | ||
| BC094236.1 | - | 7 | 701 | |||

Krr1
Genomic Map