siRNA Details
| Gene Symbol: Rufy3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001289774.1 | NP_001276703.1 | 7 | 1040 | ||
| NM_001289775.1 | NP_001276704.1 | 7 | 1040 | |||
| NM_001289776.1 | NP_001276705.1 | 8 | 1462 | |||
| NM_001289777.1 | NP_001276706.1 | 9 | 1287 | |||
| NM_027530.3 | NP_081806.1 | 7 | 1408 | |||
| GenBank mRNA | AK013185.1 | - | 9 | 1288 | ||
| AK018184.1 | - | 7 | 1386 | |||
| AK083242.1 | - | 7 | 1343 | |||
| AK148795.1 | - | 7 | 704 | |||
| AK154941.1 | - | 7 | 1041 | |||
| AK160820.1 | - | 7 | 1041 | |||
| BC017648.1 | - | 7 | 797 | 4527858 | ||
| BC049127.1 | - | 7 | 1347 | 6821567 | ||
| BC058259.1 | - | 8 | 1181 | 6469137 | ||
| CT010404.1 | - | 7 | 781 | |||

Rufy3
Genomic Map