siRNA Details
| Gene Symbol: Lair1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001113474.1 | NP_001106945.1 | 5, 6 | 670 | ||
| NM_178611.6 | NP_848726.1 | 4, 5 | 340 | |||
| GenBank mRNA | AF479685.1 | - | 5, 6 | 605 | ||
| AF491829.1 | - | 5, 6 | 703 | |||
| AF491830.1 | - | 4, 5 | 252 | |||
| AF491831.1 | - | 5, 6 | 582 | |||
| AK017222.1 | - | 4, 5 | 344 | |||
| AK033472.1 | - | 4, 5 | 317 | |||
| AK038574.1 | - | 4, 5 | 303 | |||
| AK043664.1 | - | 4, 5 | 397 | |||
| AK044465.1 | - | 5, 6 | 641 | |||
| AK050281.1 | - | 5, 6 | 626 | |||
| AK054158.1 | - | 5, 6 | 749 | |||
| AY392763.1 | - | 5, 6 | 607 | |||
| AY392764.1 | - | 4, 5 | 277 | |||
| AY392765.1 | - | 3, 4 | 160 | |||
| AY392766.1 | - | 4, 5 | 490 | |||
| BC025901.1 | - | 5, 6 | 634 | 5253846 | ||
| BY204263.1 | - | 3, 4 | 284 | |||

Lair1
Genomic Map