siRNA Details
| Gene Symbol: Mtg2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001083328.1 | NP_001076797.1 | 9 | 1173 | ||
| NM_001083329.1 | NP_001076798.1 | 8 | 1079 | |||
| NM_028422.1 | NP_082698.1 | 9 | 1218 | |||
| NM_181424.3 | NP_852089.2 | 10 | 1312 | |||
| GenBank mRNA | AK007444.1 | - | 7 | 1164 | ||
| AK040689.1 | - | 9 | 1702 | |||
| AK180040.1 | - | 2 | 169 | |||
| AK183355.1 | - | 3 | 320 | |||
| AK194297.1 | - | 2 | 170 | |||
| AK218706.1 | - | 2 | 170 | |||
| BC025144.1 | - | 10 | 1341 | 4483063 | ||
| BC028465.1 | - | 4 | 614 | |||
| BC028518.1 | - | 5 | 699 | |||
| BC030918.1 | - | 2 | 2316 | |||
| BC039784.1 | - | 2 | 2327 | |||

Mtg2
Genomic Map