siRNA Details
| Gene Symbol: D19Bwg1357e | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK050829.1 | - | 14 | 1337 | ||
| AK088081.1 | - | 14 | 1332 | |||
| AK141893.1 | - | 14 | 1335 | |||
| AK172872.1 | - | 10 | 843 | |||
| BC016186.1 | - | 14 | 1350 | |||
| BC041111.1 | - | 13 | 1315 | |||
| BC145533.1 | - | 14 | 1330 | |||

D19Bwg1357e
Genomic Map