siRNA Details
| Gene Symbol: D19Bwg1357e | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK050829.1 | - | 5, 6 | 562 | ||
| AK088081.1 | - | 5, 6 | 557 | |||
| AK141893.1 | - | 5, 6 | 560 | |||
| AK172872.1 | - | 1, 2 | 68 | |||
| BC016186.1 | - | 5, 6 | 575 | |||
| BC041111.1 | - | 4, 5 | 540 | |||
| BC052686.1 | - | 4, 5 | 513 | |||
| BC145533.1 | - | 5, 6 | 559 | |||

D19Bwg1357e
Genomic Map