siRNA Details
| Gene Symbol: Rgs7bp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029879.2 | NP_084155.2 | 4, 5 | 1335 | ||
| GenBank mRNA | AK028323.1 | - | 4, 5 | 712 | ||
| AK032138.1 | - | 4, 5 | 1335 | |||
| AK036240.1 | - | 4, 5 | 1242 | |||
| AK043529.1 | - | 1, 2 | 139 | |||
| AK046733.1 | - | 4, 5 | 964 | |||
| AK137417.1 | - | 4, 5 | 1209 | |||
| BC118957.2 | - | 4, 5 | 867 | 40093423 | ||
| DQ104214.1 | - | 4, 5 | 625 | |||

Rgs7bp
Genomic Map