siRNA Details
| Gene Symbol: Sco1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001040026.1 | NP_001035115.1 | 3 | 489 | ||
| GenBank mRNA | AK011251.1 | - | 3 | 505 | ||
| AK017794.1 | - | 3 | 490 | |||
| AK139512.1 | - | 3 | 460 | |||
| AK167305.1 | - | 3 | 354 | |||
| BC139008.1 | - | 3 | 484 | 8862030 | ||
| BC139009.1 | - | 3 | 484 | 8862031 | ||

Sco1
Genomic Map