siRNA Details
| Gene Symbol: Ahi1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026203.3 | NP_080479.2 | 11 | 2166 | ||
| GenBank mRNA | AK036336.1 | - | 11 | 2056 | ||
| AY133241.1 | - | 11 | 2059 | |||
| BC038502.1 | - | 4 | 517 | |||
| BC055400.1 | - | 10 | 1888 | 6467369 | ||
| BC065146.1 | - | 11 | 2062 | |||

Ahi1
Genomic Map