siRNA Details
| Gene Symbol: Stx7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016797.4 | NP_058077.2 | 4 | 651 | ||
| GenBank mRNA | AB019212.1 | - | 4 | 321 | ||
| AF056323.1 | - | 4 | 263 | |||
| AK002867.1 | - | 4 | 320 | |||
| AK013869.1 | - | 4 | 305 | |||
| AK082927.1 | - | 4 | 291 | |||
| AK142742.1 | - | 298 | ||||
| AK158923.1 | - | 4 | 652 | |||
| AK166822.1 | - | 4 | 321 | |||
| AK169822.1 | - | 4 | 324 | |||
| AK186903.1 | - | 2 | 40 | |||
| BC132123.1 | - | 3 | 201 | 40130400 | ||
| BC132125.1 | - | 3 | 201 | 40130402 | ||

Stx7
Genomic Map