siRNA Details
| Gene Symbol: Mtmr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016985.2 | NP_058681.1 | 10 | 1260 | ||
| GenBank mRNA | AF073997.1 | - | 10 | 1233 | ||
| AK051390.1 | - | 3 | 258 | |||
| BC056376.1 | - | 10 | 1260 | 6849757 | ||
| BC057337.1 | - | 10 | 1254 | 6825916 | ||

Mtmr1
Genomic Map