siRNA Details
| Gene Symbol: Hnrnpa2b1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016806.3 | NP_058086.2 | 4 | 676 | ||
| NM_182650.4 | NP_872591.1 | 4 | 676 | |||
| GenBank mRNA | AF073993.1 | - | 4 | 554 | ||
| AF406651.2 | - | 3 | 465 | |||
| AK049771.1 | - | 3 | 3041 | |||
| AK089012.1 | - | 4 | 627 | |||
| AK137214.1 | - | 4 | 677 | |||
| BC059107.1 | - | 2 | 212 | |||
| BC141253.1 | - | 4 | 510 | 9055865 | ||
| BC145588.1 | - | 4 | 510 | 9054154 | ||

Hnrnpa2b1
Genomic Map