siRNA Details
Gene Symbol: Txnl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016792.4 | NP_058072.2 | 1 | 262 | ||
GenBank mRNA | AF052660.1 | - | 1 | 91 | ||
AK029807.1 | - | 1 | 263 | |||
AK051698.1 | - | 1 | 237 | |||
AK144211.1 | - | 1 | 219 | |||
AK150326.1 | - | 1 | 225 | |||
AK150887.1 | - | 1 | 231 | |||
AK150983.1 | - | 1 | 231 | |||
AK152101.1 | - | 1 | 231 | |||
AK168712.1 | - | 1 | 226 | |||
BC061123.1 | - | 1 | 289 | 6596299 |
