siRNA Details
| Gene Symbol: B4galt2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001253381.1 | NP_001240310.1 | 6 | 1247 | ||
| NM_017377.5 | NP_059073.1 | 6 | 1212 | |||
| GenBank mRNA | AB019541.2 | - | 4 | 751 | ||
| AF142670.1 | - | 5 | 856 | |||
| AK145250.1 | - | 7 | 1269 | |||
| AK165829.1 | - | 6 | 1206 | |||
| BC138567.1 | - | 5 | 848 | 8861587 | ||
| BC138569.1 | - | 5 | 848 | 8861589 | ||
| BC145335.1 | - | 5 | 883 | 9053891 | ||

B4galt2
Genomic Map