siRNA Details
| Gene Symbol: Sec61a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016906.4 | NP_058602.1 | 12 | 1492 | ||
| GenBank mRNA | AB032902.1 | - | 12 | 1504 | ||
| AF145253.1 | - | 12 | 1414 | |||
| AF222743.1 | - | 12 | 1414 | |||
| AK088473.1 | - | 12 | 1493 | |||
| AK153055.1 | - | 12 | 1482 | |||
| AK159349.1 | - | 12 | 1480 | |||
| AK167484.1 | - | 12 | 1422 | |||
| BC003707.1 | - | 12 | 1418 | 3592772 | ||
| BC006020.1 | - | 8 | 1077 | |||

Sec61a1
Genomic Map