siRNA Details
| Gene Symbol: Tsnax | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016909.2 | NP_058605.1 | 5 | 639 | ||
| GenBank mRNA | AF187040.1 | - | 5 | 639 | ||
| AK014437.1 | - | 5 | 586 | |||
| AK030407.1 | - | 5 | 569 | |||
| AK140475.1 | - | 1 | 1469 | |||
| AK146339.1 | - | 5 | 572 | |||
| AK159223.1 | - | 5 | 641 | |||
| AK164453.1 | - | 4 | 414 | |||
| BC004611.1 | - | 5 | 541 | 3585400 | ||

Tsnax
Genomic Map